siRNA Details
Gene Symbol: Olr287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173334.1 | NP_775456.1 | 1 | 834 | ||
GenBank mRNA | M64378.1 | - | 1 | 834 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173334.1 | NP_775456.1 | 1 | 834 | ||
GenBank mRNA | M64378.1 | - | 1 | 834 |