siRNA Details
| Gene Symbol: Fcgr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175756.1 | NP_786932.1 | 7 | 1018 | ||
| XM_006250233.1 | XP_006250295.1 | 8 | 1176 | |||
| XM_006250234.2 | XP_006250296.1 | 9 | 1189 | |||
| XM_006250235.1 | XP_006250297.1 | 7 | 1032 | |||
| GenBank mRNA | BC091113.1 | - | 7 | 1012 | 7323350 | |
| EF424788.1 | - | 8 | 1128 | |||
| FQ228752.1 | - | 7 | 1037 | |||
| X73371.1 | - | 7 | 1052 | |||

Fcgr2b
Genomic Map