siRNA Details
| Gene Symbol: Kdm3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175764.2 | NP_786940.1 | 3, 4 | 418 | ||
| XM_006236637.2 | XP_006236699.1 | 5, 6 | 682 | |||
| XM_006236639.2 | XP_006236701.1 | 4, 5 | 639 | |||
| GenBank mRNA | FQ222223.1 | - | 4, 5 | 654 | ||
| X59993.1 | - | 3, 4 | 418 | |||

Kdm3a
Genomic Map