siRNA Details
| Gene Symbol: Abra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175844.3 | NP_787038.1 | 1 | 224 | ||
| GenBank mRNA | AF336113.1 | - | 1 | 224 | ||
| BC158575.1 | - | 1 | 210 | 9033092 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abra| Gene Symbol: Abra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175844.3 | NP_787038.1 | 1 | 224 | ||
| GenBank mRNA | AF336113.1 | - | 1 | 224 | ||
| BC158575.1 | - | 1 | 210 | 9033092 | ||