siRNA Details
Gene Symbol: C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176074.3 | NP_788263.1 | 4 | 471 | ||
GenBank mRNA | AY230250.1 | - | 4 | 473 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176074.3 | NP_788263.1 | 4 | 471 | ||
GenBank mRNA | AY230250.1 | - | 4 | 473 |