siRNA Details
| Gene Symbol: G6pc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176077.3 | NP_788266.2 | 6 | 1011 | ||
| GenBank mRNA | AY186240.1 | - | 7 | 841 | ||
| BC078689.1 | - | 6 | 1011 | 7134166 | ||
| FQ231610.1 | - | 6 | 908 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G6pc3| Gene Symbol: G6pc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176077.3 | NP_788266.2 | 6 | 1011 | ||
| GenBank mRNA | AY186240.1 | - | 7 | 841 | ||
| BC078689.1 | - | 6 | 1011 | 7134166 | ||
| FQ231610.1 | - | 6 | 908 | |||