siRNA Details
| Gene Symbol: Xrcc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177419.2 | NP_803154.1 | 3 | 255 | ||
| GenBank mRNA | BC061576.1 | - | 3 | 258 | 6887693 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xrcc5| Gene Symbol: Xrcc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177419.2 | NP_803154.1 | 3 | 255 | ||
| GenBank mRNA | BC061576.1 | - | 3 | 258 | 6887693 | |