siRNA Details
| Gene Symbol: SCG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013243.3 | NP_037375.2 | 1, 2 | 484 | ||
| GenBank mRNA | AF078851.1 | - | 1, 2 | 366 | ||
| AF453583.1 | - | 1, 2 | 489 | |||
| AK075314.1 | - | 1, 2 | 343 | |||
| AK290175.1 | - | 1, 2 | 320 | |||
| AY359093.1 | - | 1, 2 | 345 | |||
| BC014539.1 | - | 1, 2 | 120 | 4283082 | ||
| BI552493.1 | - | 1, 2 | 489 | |||

SCG3
Genomic Map