siRNA Details
| Gene Symbol: Tmem132a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178021.1 | NP_821140.1 | 4 | 817 | ||
| XM_006231041.2 | XP_006231103.1 | 4 | 1752 | |||
| GenBank mRNA | AY216677.1 | - | 4 | 817 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem132a| Gene Symbol: Tmem132a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178021.1 | NP_821140.1 | 4 | 817 | ||
| XM_006231041.2 | XP_006231103.1 | 4 | 1752 | |||
| GenBank mRNA | AY216677.1 | - | 4 | 817 | ||