siRNA Details
| Gene Symbol: Lhfpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181085.2 | NP_851599.1 | 2 | 768 | ||
| XM_008773405.1 | XP_008771627.1 | 3 | 801 | |||
| GenBank mRNA | AY253667.1 | - | 2 | 768 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lhfpl1| Gene Symbol: Lhfpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181085.2 | NP_851599.1 | 2 | 768 | ||
| XM_008773405.1 | XP_008771627.1 | 3 | 801 | |||
| GenBank mRNA | AY253667.1 | - | 2 | 768 | ||