siRNA Details
| Gene Symbol: Rhot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181823.1 | NP_861544.1 | 4 | 533 | ||
| GenBank mRNA | AY303973.1 | - | 4 | 533 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhot2| Gene Symbol: Rhot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181823.1 | NP_861544.1 | 4 | 533 | ||
| GenBank mRNA | AY303973.1 | - | 4 | 533 | ||