siRNA Details
| Gene Symbol: Chp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182738.1 | NP_877402.1 | 4 | 320 | ||
| XM_008759745.1 | XP_008757967.1 | 5 | 498 | |||
| XM_008759746.1 | XP_008757968.1 | 5 | 468 | |||
| GenBank mRNA | AB086189.1 | - | 4 | 320 | ||

Chp2
Genomic Map