siRNA Details
| Gene Symbol: Chp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182738.1 | NP_877402.1 | 5, 6 | 444 | ||
| XM_008759745.1 | XP_008757967.1 | 6, 7 | 622 | |||
| XM_008759746.1 | XP_008757968.1 | 6, 7 | 592 | |||
| GenBank mRNA | AB086189.1 | - | 5, 6 | 444 | ||

Chp2
Genomic Map