siRNA Details
| Gene Symbol: Zfp472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182823.1 | NP_877975.1 | 2 | 211 | ||
| GenBank mRNA | AY195874.1 | - | 2 | 211 | ||
| Gene Symbol: Zfp955a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC168190.1 | - | 2 | 170 | ||

Zfp472
Genomic Map