siRNA Details
| Gene Symbol: Zfp472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182823.1 | NP_877975.1 | 4 | 837 | ||
| GenBank mRNA | AY195874.1 | - | 4 | 837 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp472| Gene Symbol: Zfp472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182823.1 | NP_877975.1 | 4 | 837 | ||
| GenBank mRNA | AY195874.1 | - | 4 | 837 | ||