siRNA Details
| Gene Symbol: Stk40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183056.2 | NP_898879.2 | 11 | 3342 | ||
| XM_006238881.2 | XP_006238943.1 | 11 | 5052 | |||
| GenBank mRNA | AY336055.1 | - | 11 | 3354 | ||
| BC100062.1 | - | 11 | 3342 | 7124923 | ||

Stk40
Genomic Map