siRNA Details
| Gene Symbol: Bst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198134.2 | NP_937767.1 | 2 | 384 | ||
| GenBank mRNA | AJ538349.1 | - | 2 | 385 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bst2| Gene Symbol: Bst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198134.2 | NP_937767.1 | 2 | 384 | ||
| GenBank mRNA | AJ538349.1 | - | 2 | 385 | ||