siRNA Details
| Gene Symbol: Rdh16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199208.1 | NP_954678.1 | 1602 | |||
| GenBank mRNA | BC062000.1 | - | 1566 | 5599919 | ||
| BC079153.1 | - | 4 | 1423 | 7128686 | ||
| U33500.1 | - | 1602 | ||||
| Gene Symbol: LOC100365958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241512.2 | XP_006241574.1 | 4 | 1577 | ||

Rdh16
Genomic Map