siRNA Details
| Gene Symbol: Pcsk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199253.2 | NP_954862.2 | 11 | 2008 | ||
| GenBank mRNA | AY847775.1 | - | 11 | 2008 | ||
| BC133063.1 | - | 11 | 1939 | 8371686 | ||
| FQ219599.1 | - | 2 | 236 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcsk9| Gene Symbol: Pcsk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199253.2 | NP_954862.2 | 11 | 2008 | ||
| GenBank mRNA | AY847775.1 | - | 11 | 2008 | ||
| BC133063.1 | - | 11 | 1939 | 8371686 | ||
| FQ219599.1 | - | 2 | 236 | |||