siRNA Details
| Gene Symbol: Dstyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199463.2 | NP_955750.1 | 2 | 352 | ||
| XM_006249774.2 | XP_006249836.1 | 2 | 202 | |||
| GenBank mRNA | AY208851.1 | - | 2 | 283 | ||
| AY429675.1 | - | 2 | 352 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dstyk| Gene Symbol: Dstyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199463.2 | NP_955750.1 | 2 | 352 | ||
| XM_006249774.2 | XP_006249836.1 | 2 | 202 | |||
| GenBank mRNA | AY208851.1 | - | 2 | 283 | ||
| AY429675.1 | - | 2 | 352 | |||