siRNA Details
| Gene Symbol: ZNF180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278509.2 | NP_001265438.2 | 2, 3 | 339 | ||
| NM_001288759.2 | NP_001275688.2 | 2, 3 | 416 | |||
| NM_001288761.2 | NP_001275690.1 | 2, 3 | 339 | |||
| NM_001288762.2 | NP_001275691.1 | 2, 3 | 416 | |||
| NM_001291633.1 | NP_001278562.1 | 2, 3 | 334 | |||
| NM_013256.5 | NP_037388.3 | 2, 3 | 411 | |||
| XM_011527280.2 | XP_011525582.1 | 2, 3 | 2744 | |||
| XM_017027250.1 | XP_016882739.1 | 2, 3 | 415 | |||
| GenBank mRNA | AF192913.1 | - | 3, 4 | 393 | ||
| AU132370.1 | - | 2, 3 | 331 | |||
| DA442282.1 | - | 2, 3 | 338 | |||
| DC358291.1 | - | 2, 3 | 413 | |||

ZNF180
Genomic Map