siRNA Details
| Gene Symbol: Fut7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199491.1 | NP_955785.1 | 3 | 1825 | ||
| GenBank mRNA | AJ276068.2 | - | 3 | 1925 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fut7| Gene Symbol: Fut7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199491.1 | NP_955785.1 | 3 | 1825 | ||
| GenBank mRNA | AJ276068.2 | - | 3 | 1925 | ||