siRNA Details
| Gene Symbol: RGD735029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199493.1 | NP_955787.1 | 2 | 113 | ||
| XM_006254634.2 | XP_006254696.1 | 2 | 220 | |||
| GenBank mRNA | BC064660.1 | - | 2 | 113 | 5623815 | |
| FQ211991.1 | - | 169 | ||||

RGD735029
Genomic Map