siRNA Details
| Gene Symbol: Map1lc3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199500.2 | NP_955794.1 | 3 | 351 | ||
| GenBank mRNA | AY206668.1 | - | 3 | 299 | ||
| BC086389.1 | - | 3 | 312 | 7302206 | ||
| FM036735.1 | - | 2 | 136 | |||
| FM046244.1 | - | 3 | 319 | |||

Map1lc3a
Genomic Map