siRNA Details
| Gene Symbol: Cdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199501.1 | NP_955795.1 | 3 | 383 | ||
| XM_006240778.1 | XP_006240840.1 | 3 | 459 | |||
| GenBank mRNA | BC061832.1 | - | 3 | 383 | 5599608 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk2| Gene Symbol: Cdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199501.1 | NP_955795.1 | 3 | 383 | ||
| XM_006240778.1 | XP_006240840.1 | 3 | 459 | |||
| GenBank mRNA | BC061832.1 | - | 3 | 383 | 5599608 | |