siRNA Details
| Gene Symbol: Rxfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201417.1 | NP_958820.1 | 4 | 354 | ||
| XM_006232508.2 | XP_006232570.1 | 4 | 494 | |||
| GenBank mRNA | AY509976.1 | - | 4 | 354 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rxfp1| Gene Symbol: Rxfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201417.1 | NP_958820.1 | 4 | 354 | ||
| XM_006232508.2 | XP_006232570.1 | 4 | 494 | |||
| GenBank mRNA | AY509976.1 | - | 4 | 354 | ||