siRNA Details
Gene Symbol: Rxfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201417.1 | NP_958820.1 | 12, 13 | 968 | ||
XM_006232508.2 | XP_006232570.1 | 12, 13 | 1108 | |||
XM_006232510.2 | XP_006232572.1 | 9, 10 | 659 | |||
XM_008761085.1 | XP_008759307.1 | 8, 9 | 2409 | |||
XM_008761086.1 | XP_008759308.1 | 9, 10 | 1449 | |||
XM_008761087.1 | XP_008759309.1 | 8, 9 | 591 | |||
XM_008761088.1 | XP_008759310.1 | 8, 9 | 523 | |||
GenBank mRNA | AY509976.1 | - | 12, 13 | 968 |
