siRNA Details
| Gene Symbol: Clca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201419.1 | NP_958822.1 | 4 | 480 | ||
| GenBank mRNA | AY333961.1 | - | 4 | 480 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clca4| Gene Symbol: Clca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201419.1 | NP_958822.1 | 4 | 480 | ||
| GenBank mRNA | AY333961.1 | - | 4 | 480 | ||