siRNA Details
| Gene Symbol: Fgfr1op2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201421.1 | NP_958824.1 | 7 | 1385 | ||
| XM_006237688.2 | XP_006237750.1 | 6 | 1260 | |||
| GenBank mRNA | AY426739.1 | - | 6 | 1271 | ||
| AY426740.1 | - | 7 | 1385 | |||
| BC087696.1 | - | 6 | 1168 | 7312267 | ||
| FQ234144.1 | - | 1 | 430 | |||
| FQ235128.1 | - | 1 | 430 | |||

Fgfr1op2
Genomic Map