siRNA Details
| Gene Symbol: Tepp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201655.1 | NP_970617.1 | 8 | 947 | ||
| XM_008772274.1 | XP_008770496.1 | 7 | 1330 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tepp| Gene Symbol: Tepp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201655.1 | NP_970617.1 | 8 | 947 | ||
| XM_008772274.1 | XP_008770496.1 | 7 | 1330 | |||