siRNA Details
| Gene Symbol: Fcar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201992.1 | NP_973721.1 | 202 | |||
| GenBank mRNA | AB109766.1 | - | 202 | |||
| AB109767.1 | - | 202 | ||||
| AB109768.1 | - | 158 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fcar| Gene Symbol: Fcar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201992.1 | NP_973721.1 | 202 | |||
| GenBank mRNA | AB109766.1 | - | 202 | |||
| AB109767.1 | - | 202 | ||||
| AB109768.1 | - | 158 | ||||