siRNA Details
Gene Symbol: Fcar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201992.1 | NP_973721.1 | 486 | |||
GenBank mRNA | AB109766.1 | - | 486 | |||
AB109767.1 | - | 486 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fcar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201992.1 | NP_973721.1 | 486 | |||
GenBank mRNA | AB109766.1 | - | 486 | |||
AB109767.1 | - | 486 |