siRNA Details
| Gene Symbol: Scgb2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203333.2 | NP_976078.1 | 3 | 835 | ||
| GenBank mRNA | BC062401.1 | - | 3 | 795 | 5621455 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb2a2| Gene Symbol: Scgb2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203333.2 | NP_976078.1 | 3 | 835 | ||
| GenBank mRNA | BC062401.1 | - | 3 | 795 | 5621455 | |