siRNA Details
| Gene Symbol: Vkorc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203335.2 | NP_976080.1 | 3 | 638 | ||
| GenBank mRNA | BC166413.1 | - | 634 | 9100951 | ||
| FQ210919.1 | - | 3 | 680 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vkorc1| Gene Symbol: Vkorc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203335.2 | NP_976080.1 | 3 | 638 | ||
| GenBank mRNA | BC166413.1 | - | 634 | 9100951 | ||
| FQ210919.1 | - | 3 | 680 | |||