siRNA Details
| Gene Symbol: St3gal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203337.2 | NP_976082.1 | 3, 4 | 333 | ||
| XM_006242789.2 | XP_006242851.1 | 4, 5 | 449 | |||
| XM_008766058.1 | XP_008764280.1 | 3, 4 | 835 | |||
| XM_008766060.1 | XP_008764282.1 | 3, 4 | 1283 | |||
| GenBank mRNA | AJ626825.1 | - | 2, 3 | 97 | ||
| BC089057.1 | - | 3, 4 | 333 | 7315905 | ||

St3gal4
Genomic Map