siRNA Details
| Gene Symbol: Dmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203493.3 | NP_987089.2 | 4 | 1403 | ||
| XM_006250622.2 | XP_006250684.1 | 9 | 4021 | |||
| XM_008769994.1 | XP_008768216.1 | 5 | 1719 | |||
| XM_008769995.1 | XP_008768217.1 | 8 | 2138 | |||
| GenBank mRNA | BC129082.1 | - | 4 | 1403 | 8365946 | |
| L11354.1 | - | 5 | 1537 | |||

Dmp1
Genomic Map