siRNA Details
| Gene Symbol: Selk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207589.3 | NP_997472.2 | 4 | 303 | ||
| GenBank mRNA | AY319924.1 | - | 4 | 226 | ||
| BC088086.1 | - | 4 | 247 | 7326199 | ||
| CF979418.1 | - | 4 | 307 | |||
| FQ217552.1 | - | 2 | 161 | |||

Selk
Genomic Map