siRNA Details
| Gene Symbol: Nmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207590.1 | NP_997473.1 | 2 | 227 | ||
| GenBank mRNA | AJ629217.1 | - | 2 | 227 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nmt2| Gene Symbol: Nmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207590.1 | NP_997473.1 | 2 | 227 | ||
| GenBank mRNA | AJ629217.1 | - | 2 | 227 | ||