siRNA Details
| Gene Symbol: MAT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013283.4 | NP_037415.1 | 1, 2 | 233 | ||
| NM_182796.2 | NP_877725.1 | 1, 2 | 372 | |||
| GenBank mRNA | AB073390.1 | - | 1, 2 | 147 | ||
| AF113225.1 | - | 1, 2 | 29 | |||
| AF182814.1 | - | 1, 2 | 74 | |||
| AJ243721.1 | - | 1, 2 | 212 | |||
| AK096178.1 | - | 1, 2 | 135 | |||
| AK291919.1 | - | 1, 2 | 183 | |||
| AK294069.1 | - | 1, 2 | 45 | |||
| AK312365.1 | - | 1, 2 | 182 | |||
| AL136664.1 | - | 1, 2 | 146 | |||
| AU279489.1 | - | 1, 2 | 182 | |||
| AW966728.1 | - | 1, 2 | 211 | |||
| AY358695.1 | - | 1, 2 | 127 | |||
| BC005218.1 | - | 1, 2 | 153 | 3931953 | ||
| BC066645.1 | - | 1, 2 | 108 | 5749043 | ||
| BC093030.1 | - | 1, 2 | 142 | 30527606 | ||
| DA146198.1 | - | 1, 2 | 236 | |||
| DQ395260.1 | - | 2, 3 | 214 | |||
| DQ413183.1 | - | 1, 2 | 211 | |||
| HY091756.1 | - | 1, 2 | 373 | |||

MAT2B
Genomic Map