siRNA Details
| Gene Symbol: Chst7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207600.2 | NP_997483.1 | 1 | 779 | ||
| GenBank mRNA | AY216524.1 | - | 1 | 668 | ||
| BC111079.1 | - | 1 | 779 | 7387313 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chst7| Gene Symbol: Chst7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207600.2 | NP_997483.1 | 1 | 779 | ||
| GenBank mRNA | AY216524.1 | - | 1 | 668 | ||
| BC111079.1 | - | 1 | 779 | 7387313 | ||