siRNA Details
| Gene Symbol: Sh2d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207605.3 | NP_997488.2 | 3 | 325 | ||
| GenBank mRNA | AF468787.2 | - | 3 | 317 | ||
| BC088087.1 | - | 3 | 325 | 7379832 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh2d2a| Gene Symbol: Sh2d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207605.3 | NP_997488.2 | 3 | 325 | ||
| GenBank mRNA | AF468787.2 | - | 3 | 317 | ||
| BC088087.1 | - | 3 | 325 | 7379832 | ||