siRNA Details
| Gene Symbol: Kirrel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207606.1 | NP_997489.1 | 15 | 1909 | ||
| XM_006232675.2 | XP_006232737.1 | 16 | 2247 | |||
| XM_006232676.2 | XP_006232738.1 | 16 | 2244 | |||
| XM_006232677.2 | XP_006232739.1 | 16 | 2199 | |||
| XM_006232678.2 | XP_006232740.1 | 15 | 2101 | |||
| GenBank mRNA | AY249056.1 | - | 15 | 1909 | ||
| AY271309.1 | - | 14 | 1658 | |||

Kirrel
Genomic Map