siRNA Details
| Gene Symbol: Ns5atp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207607.1 | NP_997490.1 | 1 | 316 | ||
| GenBank mRNA | AY533136.1 | - | 1 | 316 | ||
| Gene Symbol: LOC361985 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC098673.1 | - | 4 | 472 | 7130789 | |
| BC105817.1 | - | 4 | 474 | 7368048 | ||
| FQ214790.1 | - | 4 | 507 | |||

Ns5atp4
Genomic Map