siRNA Details
| Gene Symbol: Ube4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207610.1 | NP_997493.1 | 5, 6 | 708 | ||
| XM_006242938.2 | XP_006243000.1 | 6, 7 | 922 | |||
| GenBank mRNA | AY325198.1 | - | 5, 6 | 660 | ||
| BC061761.1 | - | 5, 6 | 708 | 5621078 | ||
| FQ231663.1 | - | 2, 3 | 270 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_145400, AK028721, AK028850, BC006649, BC025043, and others.

Ube4a
Genomic Map