siRNA Details
| Gene Symbol: Ostn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207612.2 | NP_997495.1 | 2, 3 | 322 | ||
| XM_008768801.1 | XP_008767023.1 | 3, 4 | 514 | |||
| XM_008768803.1 | XP_008767025.1 | 4, 5 | 664 | |||
| GenBank mRNA | AY398682.1 | - | 324 | |||
| AY573934.1 | - | 2, 3 | 322 | |||

Ostn
Genomic Map