siRNA Details
| Gene Symbol: ST8SIA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013305.5 | NP_037437.2 | 1, 2 | 785 | ||
| XM_017025716.1 | XP_016881205.1 | 1, 2 | 426 | |||
| GenBank mRNA | AK056270.1 | - | 1, 2 | 785 | ||
| BC108910.1 | - | 1, 2 | 335 | 40011279 | ||
| BC108911.1 | - | 1, 2 | 335 | 40011281 | ||
| CR457037.1 | - | 1, 2 | 124 | |||
| U91641.1 | - | 1, 2 | 171 | |||

ST8SIA5
Genomic Map