siRNA Details
| Gene Symbol: SLC30A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321036.1 | NP_001307965.1 | 2 | 530 | ||
| NM_013309.5 | NP_037441.2 | 2 | 530 | |||
| XM_011521997.2 | XP_011520299.1 | 2 | 1034 | |||
| XM_017022560.1 | XP_016878049.1 | 1 | 336 | |||
| XM_017022561.1 | XP_016878050.1 | 2 | 528 | |||
| XM_017022562.1 | XP_016878051.1 | 1 | 336 | |||
| GenBank mRNA | AF025409.1 | - | 1 | 218 | ||
| AK290874.1 | - | 1 | 336 | |||
| BC026089.2 | - | 2 | 530 | 4829839 | ||
| BM789500.1 | - | 1 | 100 | |||

SLC30A4
Genomic Map