siRNA Details
| Gene Symbol: TAAR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003967.2 | NP_003958.2 | 1 | 1048 | ||
| GenBank mRNA | AF021818.1 | - | 1 | 996 | ||
| BC069171.1 | - | 1 | 996 | 7216924 | ||
| BC112209.1 | - | 1 | 1048 | 8327677 | ||
| BC112211.1 | - | 1 | 1048 | 8327679 | ||
More Information
Set Membership: |
|

TAAR5
Genomic Map