siRNA Details
| Gene Symbol: GPR52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005684.4 | NP_005675.3 | 1 | 1094 | ||
| GenBank mRNA | BC067456.1 | - | 1 | 1056 | 6971753 | |
| BC067457.1 | - | 1 | 1056 | 6971754 | ||
| BC067458.1 | - | 1 | 1056 | 6971755 | ||
| BC067459.1 | - | 1 | 1056 | 6971756 | ||
| BC067460.1 | - | 1 | 1056 | 6971757 | ||
| BC069460.1 | - | 1 | 1056 | 7262282 | ||
| BC095532.1 | - | 1 | 1095 | 7002024 | ||
| DA174341.1 | - | 1 | 282 | |||
| DB168074.1 | - | 1 | 128 | |||
| HQ995526.1 | - | 1 | 1056 | |||
More Information
Set Membership: |
|

GPR52
Genomic Map