siRNA Details
| Gene Symbol: SNX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199835.1 | NP_001186764.1 | 3 | 336 | ||
| NM_001199837.1 | NP_001186766.1 | 1 | 141 | |||
| NM_001318198.1 | NP_001305127.1 | 4 | 410 | |||
| NM_001318199.1 | NP_001305128.1 | 3 | 211 | |||
| NM_013322.2 | NP_037454.2 | 3 | 289 | |||
| XM_006715712.2 | XP_006715775.1 | 3 | 489 | |||
| XM_017012085.1 | XP_016867574.1 | 5 | 504 | |||
| XM_017012086.1 | XP_016867575.1 | 5 | 943 | |||
| XM_017012087.1 | XP_016867576.1 | 4 | 433 | |||
| XM_017012088.1 | XP_016867577.1 | 4 | 380 | |||
| GenBank mRNA | AF121860.1 | - | 3 | 202 | ||
| AK296142.1 | - | 4 | 240 | |||
| AK307905.1 | - | 1 | 90 | |||
| AK312850.1 | - | 3 | 211 | |||
| AK316088.1 | - | 4 | 386 | |||
| BC031050.1 | - | 3 | 183 | 5278015 | ||
| BC034992.1 | - | 3 | 262 | 4827599 | ||
| BJ995940.1 | - | 1 | 141 | |||
| BM147978.1 | - | 3 | 330 | |||
| DA377368.1 | - | 3 | 211 | |||

SNX10
Genomic Map